Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGDIB
Entrez Gene ID
327676
Gene Name
Rho GDP dissociation inhibitor beta
Gene Aliases
-
Chromosome Location
Chr.5: 94946440 - 94965335 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175797.2 NP_786991.1 4 - 5 385 150
GenBank mRNA AF182001.1 - 4 - 5 385 150
BC102109.1 - 4 - 5 407 150

View Full Product Details