Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP6AP1
Entrez Gene ID
327687
Gene Name
ATPase H+ transporting accessory protein 1
Gene Aliases
ATP6IP1, Ac45
Chromosome Location
Chr.X: 37489007 - 37496787 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175806.2 NP_787000.1 6 - 7 709 85
GenBank mRNA BC104511.1 - 6 - 7 677 85
U10039.1 - 6 - 7 709 85

View Full Product Details