Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK1A
Entrez Gene ID
282112
Gene Name
uroplakin 1A
Gene Aliases
-
Chromosome Location
Chr.18: 46395906 - 46405039 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176611.2 NP_788784.1 2 - 3 302 82
GenBank mRNA Z29475.1 - - 302 82

View Full Product Details