Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAG15
Entrez Gene ID
337909
Gene Name
pregnancy-associated glycoprotein 15
Gene Aliases
-
Chromosome Location
Chr.29: 38083591 - 38092452 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176624.1 NP_788797.1 7 - 8 889 60
GenBank mRNA AF192332.1 - 7 - 8 889 60

View Full Product Details