The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_176851.1 | NP_789821.1 | 2 - 3 | 170 | 85 |
GenBank mRNA | AJ132773.2 | - | 2 - 3 | 170 | 85 |