Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAT
Entrez Gene ID
280801
Gene Name
glycine-N-acyltransferase-like
Gene Aliases
-
Chromosome Location
Chr.15: 82088581 - 82118181 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177486.2 NP_803452.1 5 - 6 829 88
GenBank mRNA AF022073.1 - 5 - 6 829 88

View Full Product Details