Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QPCT
Entrez Gene ID
281437
Gene Name
glutaminyl-peptide cyclotransferase
Gene Aliases
-
Chromosome Location
Chr.11: 19792187 - 19822477 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177506.2 NP_803472.1 6 - 7 1032 88
GenBank mRNA BC120315.1 - 6 - 7 951 88
M80626.1 - 6 - 7 1032 88

View Full Product Details