Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDN1
Entrez Gene ID
281137
Gene Name
endothelin 1
Gene Aliases
-
Chromosome Location
Chr.23: 44156440 - 44163423 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181010.2 NP_851353.1 4 - 5 800 122
GenBank mRNA BC102459.1 - 4 - 5 820 122
S37093.1 - 4 - 5 800 122
X52740.1 - - 550 122
X52942.1 - - 662 122

View Full Product Details