Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLCA3
Entrez Gene ID
281694
Gene Name
chloride channel regulator 3
Gene Aliases
-
Chromosome Location
Chr.3: 57420369 - 57445306 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181018.2 NP_851361.1 10 - 11 1531 86
GenBank mRNA AF001261.1 - 10 - 11 1531 86
AF001262.1 - 10 - 11 1527 86
AF001263.1 - 10 - 11 1662 86

View Full Product Details