The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203363.1 | NP_976239.1 | 1 - 2 | 187 | 77 |
GenBank mRNA | AY192437.1 | - | 1 - 2 | 187 | 77 |
BC103120.1 | - | 1 - 2 | 207 | 77 |