Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGO2
Entrez Gene ID
404130
Gene Name
argonaute RISC catalytic component 2
Gene Aliases
EIF2C2
Chromosome Location
Chr.14: 3100810 - 3138228 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205794.1 NP_991363.1 12 - 13 1596 76
GenBank mRNA AY533678.1 - 12 - 13 1596 76
BC151491.1 - 12 - 13 1704 76

View Full Product Details