Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAPTM4B
Entrez Gene ID
404155
Gene Name
lysosomal protein transmembrane 4 beta
Gene Aliases
-
Chromosome Location
Chr.14: 66373386 - 66438425 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205802.2 NP_991371.2 6 - 7 844 82
GenBank mRNA AF276819.1 - 6 - 7 82
BC149605.1 - 6 - 7 842 82
EV638847.1 - 6 - 7 553 82

View Full Product Details