Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CL43
Entrez Gene ID
431784
Gene Name
collectin-43
Gene Aliases
CL-43
Chromosome Location
Chr.28: 35510715 - 35519246 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002237.1 NP_001002237.1 5 - 6 658 88
GenBank mRNA AY071821.1 - 5 - 6 658 88
BC116147.1 - 5 - 6 88
X75912.1 - - 427 88

View Full Product Details