Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLRJ1
Entrez Gene ID
444861
Gene Name
killer cell lectin-like receptor family J member 1
Gene Aliases
KLRJ, Ly49
Chromosome Location
Chr.5: 99425372 - 99435646 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002884.1 NP_001002884.1 5 - 6 518 110
GenBank mRNA AY156928.1 - 5 - 6 518 110
BC149263.1 - 5 - 6 584 110

View Full Product Details