Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLRD1
Entrez Gene ID
444877
Gene Name
killer cell lectin-like receptor subfamily D, member 1
Gene Aliases
CD94, CD94-1
Chromosome Location
Chr.5: 99623587 - 99660633 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002890.1 NP_001002890.1 1 - 2 61 66
GenBank mRNA AF422180.1 - 1 - 2 61 66

View Full Product Details