Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCAR
Entrez Gene ID
503555
Gene Name
Fc fragment of IgA receptor
Gene Aliases
-
Chromosome Location
Chr.18: 62527260 - 62539102 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012685.1 NP_001012703.1 2 - 3 69 81
GenBank mRNA AY247821.1 - 2 - 3 69 81

View Full Product Details