Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AK1
Entrez Gene ID
280715
Gene Name
adenylate kinase 1
Gene Aliases
-
Chromosome Location
Chr.11: 98538949 - 98555122 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013582.1 NP_001013600.1 6 - 7 586 70
GenBank mRNA BC114796.1 - 6 - 7 626 70
BT020951.1 - 6 - 7 586 70

View Full Product Details