Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN10
Entrez Gene ID
506545
Gene Name
claudin 10
Gene Aliases
-
Chromosome Location
Chr.12: 72904648 - 73009452 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014857.1 NP_001014857.1 3 - 4 470 96
NM_001105609.2 NP_001099079.1 3 - 4 679 96
GenBank mRNA BC153270.1 - 3 - 4 680 96
BT020910.1 - 3 - 4 470 96

View Full Product Details