Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF34
Entrez Gene ID
506764
Gene Name
ring finger protein 34
Gene Aliases
-
Chromosome Location
Chr.17: 53771759 - 53798675 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014858.1 NP_001014858.1 4 - 5 820 151
GenBank mRNA BC110246.1 - 5 - 6 1074 151
BT020924.1 - 4 - 5 820 151

View Full Product Details