Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDIP1
Entrez Gene ID
510242
Gene Name
cell death inducing p53 target 1
Gene Aliases
C16orf5, C25H16orf5
Chromosome Location
Chr.25: 3654420 - 3678820 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014892.1 NP_001014892.1 2 - 3 262 127
GenBank mRNA BC102294.1 - 2 - 3 175 127
BT021752.1 - 2 - 3 262 127

View Full Product Details