Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNL4B
Entrez Gene ID
510719
Gene Name
thioredoxin like 4B
Gene Aliases
-
Chromosome Location
Chr.18: 39027943 - 39035340 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014897.2 NP_001014897.1 1 - 2 217 98
GenBank mRNA BC103293.1 - 1 - 2 217 98
BT020765.1 - 1 - 2 217 98

View Full Product Details