Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSPBAP1
Entrez Gene ID
513211
Gene Name
HSPB1 associated protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 67064196 - 67121243 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014911.1 NP_001014911.1 7 - 8 1032 92
GenBank mRNA BT021854.1 - 7 - 8 1032 92

View Full Product Details