Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMOX1
Entrez Gene ID
513221
Gene Name
heme oxygenase 1
Gene Aliases
HO-1
Chromosome Location
Chr.5: 73623470 - 73630535 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014912.1 NP_001014912.1 2 - 3 236 105
GenBank mRNA BC102105.1 - 2 - 3 254 105
BT020968.1 - 2 - 3 236 105

View Full Product Details