Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM79
Entrez Gene ID
513599
Gene Name
transmembrane protein 79
Gene Aliases
-
Chromosome Location
Chr.3: 14509526 - 14515573 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014920.1 NP_001014920.1 3 - 4 1104 64
GenBank mRNA BC123830.1 - 2 - 3 1025 64
BT020827.1 - 3 - 4 1104 64

View Full Product Details