Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EEF1B2
Entrez Gene ID
520875
Gene Name
eukaryotic translation elongation factor 1 beta 2
Gene Aliases
EEF1B
Chromosome Location
Chr.2: 94490099 - 94493382 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014936.1 NP_001014936.1 4 - 5 424 129
GenBank mRNA AY911340.1 - 3 - 4 420 129
BC102548.1 - 3 - 4 345 129
BC147944.1 - 3 - 4 460 129
BT021037.1 - 4 - 5 424 129

View Full Product Details