Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBCB
Entrez Gene ID
530447
Gene Name
tubulin folding cofactor B
Gene Aliases
CKAP1
Chromosome Location
Chr.18: 46775238 - 46783614 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014939.1 NP_001014939.1 5 - 6 658 67
GenBank mRNA BC102571.1 - 5 - 6 67
BT021069.1 - 5 - 6 658 67

View Full Product Details