Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMAN2L
Entrez Gene ID
539289
Gene Name
lectin, mannose binding 2 like
Gene Aliases
-
Chromosome Location
Chr.11: 2683655 - 2704108 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014953.1 NP_001014953.1 5 - 6 549 76
GenBank mRNA BT021149.1 - 5 - 6 549 76

View Full Product Details