Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRR5L
Entrez Gene ID
505048
Gene Name
proline rich 5 like
Gene Aliases
PROTOR2
Chromosome Location
Chr.15: 66763192 - 66918835 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015519.1 NP_001015519.1 1 - 2 140 104
GenBank mRNA BC151482.1 - 1 - 2 131 104
BT020860.1 - 1 - 2 140 104

View Full Product Details