Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXOSC1
Entrez Gene ID
506018
Gene Name
exosome component 1
Gene Aliases
-
Chromosome Location
Chr.26: 18657376 - 18664127 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015529.1 NP_001015529.1 6 - 7 401 113
GenBank mRNA BC133440.1 - 6 - 7 418 113
BT020971.1 - 6 - 7 401 113

View Full Product Details