Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC12
Entrez Gene ID
506991
Gene Name
thioredoxin domain containing 12
Gene Aliases
ERP18, TLP19
Chromosome Location
Chr.3: 94278884 - 94312525 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015536.1 NP_001015536.1 2 - 3 264 77
GenBank mRNA BC102314.1 - 2 - 3 293 77
BT021084.1 - 2 - 3 264 77

View Full Product Details