Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC12
Entrez Gene ID
506991
Gene Name
thioredoxin domain containing 12
Gene Aliases
ERP18, TLP19
Chromosome Location
Chr.3: 94278884 - 94312525 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015536.1 NP_001015536.1 4 - 5 388 138
GenBank mRNA BC102314.1 - 4 - 5 417 138
BT021084.1 - 4 - 5 388 138

View Full Product Details