Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCAS2
Entrez Gene ID
507944
Gene Name
BCAS2 pre-mRNA processing factor
Gene Aliases
-
Chromosome Location
Chr.3: 28755553 - 28769812 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015540.1 NP_001015540.1 4 - 5 467 73
GenBank mRNA BC102652.1 - 4 - 5 487 73
BT020986.1 - 4 - 5 467 73

View Full Product Details