Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL13
Entrez Gene ID
508221
Gene Name
ribosomal protein L13
Gene Aliases
-
Chromosome Location
Chr.18: 14490266 - 14492655 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015543.2 NP_001015543.1 4 - 5 460 92
GenBank mRNA AY911336.1 - 4 - 5 461 92
BC102865.1 - 4 - 5 464 92

View Full Product Details