Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RASGEF1A
Entrez Gene ID
514877
Gene Name
RasGEF domain family member 1A
Gene Aliases
-
Chromosome Location
Chr.28: 13591898 - 13683152 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015601.1 NP_001015601.1 11 - 12 1360 60
GenBank mRNA BT020843.1 - 11 - 12 1360 60

View Full Product Details