Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RASGEF1A
Entrez Gene ID
514877
Gene Name
RasGEF domain family member 1A
Gene Aliases
-
Chromosome Location
Chr.28: 13591898 - 13683152 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015601.1 NP_001015601.1 2 - 3 259 132
GenBank mRNA BT020843.1 - 2 - 3 259 132

View Full Product Details