Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM10
Entrez Gene ID
515628
Gene Name
tripartite motif containing 10
Gene Aliases
-
Chromosome Location
Chr.23: 28816473 - 28825061 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015607.1 NP_001015607.1 1 - 2 123 107
GenBank mRNA BC114866.1 - 1 - 2 142 107
BT020956.1 - 1 - 2 123 107

View Full Product Details