Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYW3
Entrez Gene ID
519731
Gene Name
tRNA-yW synthesizing protein 3 homolog
Gene Aliases
-
Chromosome Location
Chr.3: 69968112 - 69990582 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015620.1 NP_001015620.1 4 - 5 464 103
GenBank mRNA BC114025.1 - 4 - 5 463 103
BT020826.1 - 4 - 5 464 103

View Full Product Details