The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015626.1 | NP_001015626.1 | 1 - 2 | 193 | 55 |
GenBank mRNA | BC103161.1 | - | 1 - 2 | 209 | 55 |
BT021070.1 | - | 1 - 2 | 193 | 55 |