Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS72
Entrez Gene ID
534883
Gene Name
vacuolar protein sorting 72 homolog
Gene Aliases
TCFL1
Chromosome Location
Chr.3: 19624886 - 19636725 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015658.1 NP_001015658.1 3 - 4 444 80
GenBank mRNA BC109639.1 - 3 - 4 409 80
BT020964.1 - 3 - 4 444 80

View Full Product Details