Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BRINP1
Entrez Gene ID
538990
Gene Name
BMP/retinoic acid inducible neural specific 1
Gene Aliases
DBC1
Chromosome Location
Chr.8: 108637081 - 108834345 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015669.1 NP_001015669.1 6 - 7 1290 63
GenBank mRNA BT020908.1 - 6 - 7 1290 63

View Full Product Details