Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOZ2
Entrez Gene ID
540487
Gene Name
myozenin 2
Gene Aliases
CS-1
Chromosome Location
Chr.6: 6097584 - 6136039 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015676.1 NP_001015676.1 3 - 4 381 147
GenBank mRNA BC103121.1 - 3 - 4 382 147
BT020817.1 - 3 - 4 381 147

View Full Product Details