Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM54
Entrez Gene ID
535320
Gene Name
tripartite motif containing 54
Gene Aliases
-
Chromosome Location
Chr.11: 72337035 - 72358693 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017951.1 NP_001017951.1 3 - 4 853 88
GenBank mRNA BC114171.1 - 3 - 4 858 88
BT021782.1 - 3 - 4 853 88

View Full Product Details