Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LSM1
Entrez Gene ID
535447
Gene Name
LSM1 homolog, mRNA degradation associated
Gene Aliases
-
Chromosome Location
Chr.27: 33338510 - 33348718 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017952.1 NP_001017952.1 1 - 2 211 123
GenBank mRNA BC110194.1 - 1 - 2 207 123
BT020738.1 - 1 - 2 131 123
BT020771.1 - 1 - 2 219 123
BT020779.1 - 1 - 2 211 123

View Full Product Details