Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GMFG
Entrez Gene ID
514407
Gene Name
glia maturation factor gamma
Gene Aliases
-
Chromosome Location
Chr.18: 49038392 - 49043303 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024537.2 NP_001019708.1 4 - 5 258 106
GenBank mRNA AY911328.1 - 4 - 5 259 106
BC103101.1 - 4 - 5 250 106

View Full Product Details