Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFIC
Entrez Gene ID
553098
Gene Name
nuclear factor I C
Gene Aliases
-
Chromosome Location
Chr.7: 20444934 - 20516202 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024574.1 NP_001019745.1 3 - 4 629 61
GenBank mRNA BC114080.1 - 3 - 4 702 61
BT026541.1 - 3 - 4 653 61
BT030543.1 - 2 - 3 595 61
DQ004950.1 - 3 - 4 629 61

View Full Product Details