Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS28
Entrez Gene ID
282877
Gene Name
ribosomal protein S28
Gene Aliases
-
Chromosome Location
Chr.7: 16968061 - 16969193 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025316.2 NP_001020487.1 3 - 4 255 57
GenBank mRNA AY911353.1 - 3 - 4 256 57
BC102565.1 - 1 - 1 257 57

View Full Product Details