Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC7A
Entrez Gene ID
532865
Gene Name
C-type lectin domain containing 7A
Gene Aliases
-
Chromosome Location
Chr.5: 99840395 - 99855696 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031852.1 NP_001027022.1 3 - 4 353 116
GenBank mRNA AY937383.1 - 3 - 4 353 116

View Full Product Details