Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECI2
Entrez Gene ID
505355
Gene Name
enoyl-CoA delta isomerase 2
Gene Aliases
PECI
Chromosome Location
Chr.23: 50039735 - 50057698 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC102906.1 - 5 - 6 624 86

View Full Product Details