Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPAR5
Entrez Gene ID
507598
Gene Name
lysophosphatidic acid receptor 5
Gene Aliases
GPR92
Chromosome Location
Chr.5: 103795807 - 103806563 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034304.2 NP_001029476.1 1 - 2 53 60
GenBank mRNA BC102849.1 - 1 - 2 54 60

View Full Product Details