The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034319.2 | NP_001029491.1 | 8 - 9 | 911 | 73 |
GenBank mRNA | BC102927.1 | - | 8 - 9 | 912 | 73 |
EF576938.1 | - | 8 - 9 | 896 | 73 |